Skip to Content
Merck
All Photos(1)

Key Documents

02380595

Ectoine

primary reference standard

Synonym(s):

(S)-2-Methyl-1,4,5,6-tetrahydropyrimidine-4-carboxylic acid, Thp(B)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€299.00

€299.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€299.00

About This Item

Empirical Formula (Hill Notation):
C6H10N2O2
CAS Number:
Molecular Weight:
142.16
Beilstein:
7288977
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.24

€299.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

CC1=N[C@@H](CCN1)C(O)=O

InChI

1S/C6H10N2O2/c1-4-7-3-2-5(8-4)6(9)10/h5H,2-3H2,1H3,(H,7,8)(H,9,10)/t5-/m0/s1

InChI key

WQXNXVUDBPYKBA-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service