Skip to Content
Merck
All Photos(2)

Key Documents

01913

Supelco

2-Ethoxyethyl acetate

certified reference material, TraceCERT®

Synonym(s):

1-Acetoxy-2-ethoxyethane, Cellosolve® acetate, Ethylene glycol monoethyl ether acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2OC2H5
CAS Number:
Molecular Weight:
132.16
Beilstein:
1748677
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor density

4.6 (vs air)

vapor pressure

2 mmHg ( 20 °C)

product line

TraceCERT®

autoignition temp.

715 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

13 %

storage condition

under inert gas

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.406 (lit.)

bp

156 °C (lit.)

mp

−61 °C (lit.)

density

0.975 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCOCCOC(C)=O

InChI

1S/C6H12O3/c1-3-8-4-5-9-6(2)7/h3-5H2,1-2H3

InChI key

SVONRAPFKPVNKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethoxyethyl acetate belongs to ethylene glycol ether class of compounds. It is highly miscible with water and organic solvents. It is mostly used in dyes, coatings, insecticides, soaps and cosmetics. It also has teratogenic effect and is capable to bring about testicular changes in rodents.[1]
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Cellosolve is a registered trademark of Union Carbide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L M Sweeney et al.
Toxicological sciences : an official journal of the Society of Toxicology, 62(1), 124-139 (2001-06-16)
Methoxyethanol (ethylene glycol monomethyl ether, EGME), ethoxyethanol (ethylene glycol monoethyl ether, EGEE), and ethoxyethyl acetate (ethylene glycol monoethyl ether acetate, EGEEA) are all developmental toxicants in laboratory animals. Due to the imprecise nature of the exposure data in epidemiology studies
Reproductive toxicity of ethylene glycol monomethyl ether, ethylene glycol monoethyl ether and their acetates.
J A Wess
Scandinavian journal of work, environment & health, 18 Suppl 2, 43-45 (1992-01-01)
J E Doe
Environmental health perspectives, 57, 33-41 (1984-08-01)
An inhalation teratology study was conducted in rats at 10, 50 and 250 ppm ethylene glycol monoethyl ether (EGEE) and in rabbits at 10, 50 and 175 ppm EGEE. This study was designed to supplement a study conducted for NIOSH
S E Chia et al.
American journal of industrial medicine, 31(2), 148-152 (1997-02-01)
The objectives of this study were to determine the exposure levels among workers who handle 2-ethoxyethylacetate (EGEEA) in the liquid crystal display (LCD) manufacturing industry and to study the menstrual patterns among the exposed workers compared to a referent group
Y Kim et al.
Occupational and environmental medicine, 56(6), 378-382 (1999-09-04)
To evaluate exposure to mixed solvents containing ethylene glycol monoethyl ether acetate (EGEEA) in shipyard painters, to determine if EGEEA is toxic to the bone marrow. An industrial hygiene survey was performed to identify exposure to EGEEA of two groups

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service