Skip to Content
Merck
All Photos(1)

Key Documents

01490590

Ginkgolide C

primary reference standard

Synonym(s):

1,7-Dihydroxy-ginkgolide A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O11
CAS Number:
Molecular Weight:
440.40
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

storage temp.

−20°C

SMILES string

[H][C@@]12OC(=O)[C@@H](C)[C@]1(O)[C@@]34OC5OC(=O)[C@H](O)C56[C@@H]([C@H](O)[C@@H](OC3=O)[C@@]46[C@H]2O)C(C)(C)C

InChI

1S/C20H24O11/c1-5-12(24)28-11-8(22)18-10-6(21)7(16(2,3)4)17(18)9(23)13(25)30-15(17)31-20(18,14(26)29-10)19(5,11)27/h5-11,15,21-23,27H,1-4H3/t5-,6+,7+,8+,9+,10-,11+,15?,17?,18-,19-,20-/m1/s1

InChI key

AMOGMTLMADGEOQ-CFTQXCTRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Cynarin phyproof® Reference Substance

PHL89179

Cynarin

Haixia Lin et al.
Toxicological sciences : an official journal of the Society of Toxicology, 139(2), 338-349 (2014-03-07)
Ginkgo biloba has been used for many thousand years as a traditional herbal remedy and its extract has been consumed for many decades as a dietary supplement. Ginkgo biloba leaf extract is a complex mixture with many constituents, including flavonol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service