Skip to Content
Merck
All Photos(1)

Key Documents

00793

Sigma-Aldrich

Magnesium formate dihydrate

BioXtra, ≥98.0% (RT)

Synonym(s):

Formic acid magnesium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€54.90
500 G
€217.00

€54.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€54.90
500 G
€217.00

About This Item

Empirical Formula (Hill Notation):
C2H2MgO4 · 2H2O
CAS Number:
Molecular Weight:
150.37
Beilstein:
3725251
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
powder or crystals

€54.90


Please contact Customer Service for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥98.0% (RT)

form

powder or crystals

anion traces

chloride (Cl-): ≤1000 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

Ca: ≤5000 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤500 mg/kg
Na: ≤5000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Mg++].[O-]C=O.[O-]C=O

InChI

1S/2CH2O2.Mg/c2*2-1-3;/h2*1H,(H,2,3);/q;;+2/p-2

InChI key

GMDNUWQNDQDBNQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium formate dehydrate, or formic acid magnesium salt, has been used in studies to develop a multiresidue method for determining pesticides in fruits and vegetables, as well as for the simultaneous determination of tropane alkaloids and glycoalkaloids in grains and seeds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Z Jandrić et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 28(9), 1205-1219 (2011-08-16)
A new, rapid and sensitive multiresidue method is reported for the simultaneous determination of tropane alkaloids (tropine, atropine, scopolamine, homatropine, anisodamine) and glycoalkaloids (α-solanine, α-chaconine) in grains and seeds (wheat, rye, maize, soybean, linseed). Dispersive solid phase extraction (DSPE) was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service