Skip to Content
Merck
All Photos(1)

Key Documents

00484

Sigma-Aldrich

Krypton

≥99.998%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Kr
CAS Number:
Molecular Weight:
83.80
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.998%

SMILES string

[Kr]

InChI

1S/Kr

InChI key

DNNSSWSSYDEUBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin
filling pressure at 15°C: 11 bar; content (15°C, 1 bar): 11 l

Other Notes

Sales restrictions may apply

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anne-Lise Hachulla et al.
Radiology, 263(1), 253-259 (2012-02-15)
To evaluate the tolerance and level of enhancement achievable after inhalation of stable krypton. This study was approved by the institutional review board and the local ethics committee. Written informed consent was obtained from all subjects. The study was planned
Carlos A Fernandez et al.
Journal of the American Chemical Society, 134(22), 9046-9049 (2012-05-18)
Krypton (Kr) and xenon (Xe) adsorption on two partially fluorinated metal-organic frameworks (FMOFCu and FMOFZn) with different cavity size and topologies are reported. FMOFCu shows an inversion in sorption selectivity toward Kr at temperatures below 0 °C while FMOFZn does
Dieter Hauchecorne et al.
Physical chemistry chemical physics : PCCP, 13(21), 10204-10213 (2011-04-22)
Using FTIR and Raman spectroscopy, the formation of halogen bonded complexes of the trifluorohalomethanes CF(3)Cl, CF(3)Br and CF(3)I with dimethyl sulfide (DMS) dissolved in liquid krypton has been investigated. For CF(3)Br and CF(3)I, evidence was found for the formation of
Kenta Mizuse et al.
Physical chemistry chemical physics : PCCP, 13(15), 7129-7135 (2011-03-15)
Although messenger mediated spectroscopy is a widely-used technique to study gas phase ionic species, effects of messengers themselves are not necessarily clear. In this study, we report infrared photodissociation spectroscopy of H(+)(H(2)O)(6)·M(m) (M = Ne, Ar, Kr, Xe, H(2), N(2)
Lu Zhang et al.
Optics express, 19(25), 25812-25822 (2012-01-26)
We observed that increasing the clusters size and laser pulse contrast can enhance the X-ray flux emitted by femtosecond-laser-driven-cluster plasma. By focusing a high contrast laser (10(-10)) on large argon clusters, high flux Kα-like X-rays (around 2.96 keV) is generated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service