Skip to Content
Merck
All Photos(1)

Key Documents

00043

Sigma-Aldrich

Boc-N-Me-Tyr(Bzl)-OH

≥98.0% (HPLC)

Synonym(s):

Nα-Boc-O-benzyl-Nα-methyl-L-tyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27NO5
CAS Number:
Molecular Weight:
385.45
Beilstein:
2913415
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

crystalline

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

beige

application(s)

peptide synthesis

SMILES string

CN([C@@H](Cc1ccc(OCc2ccccc2)cc1)C(O)=O)C(=O)OC(C)(C)C

InChI

1S/C22H27NO5/c1-22(2,3)28-21(26)23(4)19(20(24)25)14-16-10-12-18(13-11-16)27-15-17-8-6-5-7-9-17/h5-13,19H,14-15H2,1-4H3,(H,24,25)/t19-/m0/s1

InChI key

FSNRGORPOYPIJC-IBGZPJMESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service