Skip to Content
Merck
All Photos(1)

Key Documents

30-5450

Sigma-Aldrich

TWEEN® 20

SAJ first grade

Synonym(s):

Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
Pricing and availability is not currently available.

grade

SAJ first grade

description

non-ionic detergent

form

liquid

mol wt

~1228

availability

available only in Japan

refractive index

n20/D 1.468 (lit.)

density

1.095 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic detergent

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

566.6 °F

Flash Point(C)

297 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lycias Zembe et al.
PloS one, 6(10), e26096-e26096 (2011-10-25)
The genetic diversity of HIV-1 across the globe is a major challenge for developing an HIV vaccine. To facilitate immunogen design, it is important to characterize clusters of commonly targeted T-cell epitopes across different HIV clades. To address this, we
Daniel Bushey et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(7), 1948-1961 (2009-02-21)
Sleep need is affected by developmental stage and neuronal plasticity, but the underlying mechanisms remain unclear. The fragile X mental retardation gene Fmr1, whose loss-of-function mutation causes the most common form of inherited mental retardation in humans, is involved in
Aida Maddahi et al.
BMC neuroscience, 9, 85-85 (2008-09-17)
MEK1/2 is a serine/threonine protein that phosphorylates extracellular signal-regulated kinase (ERK1/2). Cerebral ischemia results in enhanced expression of cerebrovascular contractile receptors in the middle cerebral artery (MCA) leading to the ischemic region. Here we explored the role of the MEK/ERK
Louis Hermo et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 56(12), 1121-1135 (2008-09-18)
Water content within the epididymis of the male reproductive system is stringently regulated to promote sperm maturation. Several members of the aquaporin (AQP) family of water channel-forming integral membrane proteins have been identified in epididymal cells, but expression profiling for
Qing-wen Chen et al.
BMC cell biology, 10, 52-52 (2009-07-07)
muscle cells (VSMCs) through activation of endothelin type A (ETA) and type B (ETB) receptors. The extracellular signal-regulated kinase 1 and 2 (ERK1/2) mitogen-activated protein kinases (MAPK) are involved in ET-1-induced VSMC contraction and proliferation. This study was designed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service