Skip to Content
Merck
All Photos(1)

Key Documents

30-3905

Sigma-Aldrich

Trichloroacetic acid

≥99.5%

Synonym(s):

TCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
Beilstein:
970119
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

grade

reagent

vapor density

<1 (vs air)

vapor pressure

1 mmHg ( 51 °C)

Assay

≥99.5%

form

solid

availability

available only in Japan

refractive index

n20/D 1.62 (lit.)

bp

196 °C (lit.)

mp

54-58 °C (lit.)

density

1.62 g/mL at 25 °C (lit.)

SMILES string

OC(=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.

Suitability

for biological purposes

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Mortensen et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 18(1), 55-62 (2012-10-17)
The ratio of the nitric oxide synthase (NOS) cofactor tetrahydrobiopterin (BH(4)) to its oxidized form dihydrobiopterin (BH(2)) has been suggested as an index of endothelial dysfunction. Consequently, much effort has been put into preserving the in vivo equilibrium between these
Hikmat Al-Ahmadie et al.
Cancer discovery, 4(9), 1014-1021 (2014-06-18)
Metastatic solid tumors are almost invariably fatal. Patients with disseminated small-cell cancers have a particularly unfavorable prognosis, with most succumbing to their disease within two years. Here, we report on the genetic and functional analysis of an outlier curative response
Inga Müller et al.
PloS one, 9(5), e97285-e97285 (2014-05-27)
Members of the MYC family are the most frequently deregulated oncogenes in human cancer and are often correlated with aggressive disease and/or poorly differentiated tumors. Since patients with MYCN-amplified neuroblastoma have a poor prognosis, targeting MYCN using small molecule inhibitors
Valentina Folgiero et al.
Oncotarget, 5(8), 2052-2064 (2014-06-07)
Microenvironmental factors contribute to the immune dysfunction characterizing acute myeloid leukemia (AML). Indoleamine 2,3-dioxygenase 1 (IDO1) is an interferon (IFN)-γ-inducible enzyme that degrades tryptophan into kynurenine, which, in turn, inhibits effector T cells and promotes regulatory T-cell (Treg) differentiation. It
Chih-Ho Lai et al.
Infection and immunity, 76(7), 3293-3303 (2008-04-30)
Infection with Helicobacter pylori cagA-positive strains is associated with gastritis, ulcerations, and gastric cancer. CagA is translocated into infected epithelial cells by a type IV secretion system and can be tyrosine phosphorylated, inducing signal transduction and motogenic responses in epithelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service