Skip to Content
Merck
All Photos(1)

Key Documents

30-1850

Sigma-Aldrich

Thionyl chloride

SAJ first grade, ≥90.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SOCl2
CAS Number:
Molecular Weight:
118.97
Beilstein:
1209273
MDL number:
UNSPSC Code:
12352307
PubChem Substance ID:

grade

SAJ first grade

vapor pressure

97 mmHg ( 20 °C)

Assay

≥90.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.518 (lit.)

bp

79 °C (lit.)

mp

−105 °C (lit.)

density

1.631 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

ClS(Cl)=O

InChI

1S/Cl2OS/c1-4(2)3

InChI key

FYSNRJHAOHDILO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gwenaël Chamoulaud et al.
Langmuir : the ACS journal of surfaces and colloids, 20(12), 4989-4995 (2005-06-30)
This paper describes a novel approach for the surface modification of a cation-exchange membrane, bearing sulfonate groups, by a cationic layer. The modification procedure involved the chlorosulfonation of the sulfonate groups of the base membrane with thionyl chloride, followed by
Feng Xu et al.
The Journal of organic chemistry, 73(1), 312-315 (2007-12-07)
A simple, one-pot preparation of cyclic amines via efficient chlorination of amino alcohols with use of SOCl(2) has been developed. This approach obviates the need for the classical N-protection/O-activation/cyclization/deprotection sequence commonly employed for this type of transformation. The reaction pathways
Alexei Chichinin et al.
Physical chemistry chemical physics : PCCP, 7(2), 301-309 (2005-01-21)
New theoretical and experimental results for the ultraviolet photodissociation dynamics of thionyl chloride (SOCl2) are presented and combined with existing data from a variety of sources in order to provide a unified view of the photodissociation dynamics of SOC12. Time-dependent
Hend N Hafez et al.
Bioorganic & medicinal chemistry letters, 19(15), 4143-4147 (2009-06-23)
Chlorosulfonation of 3-methyl[1,2,4]triazolo[4,3-a]pyrimidine with chlorosulfonic acid in the presence of thionyl chloride was studied. When triazolo[4,3-a]pyrimidines are used as substrates, the substitution occurs at C-6. Also the reactivity of the hydrazides (7) towards aldehydes, thioglycolic acid and amines were studies.
S Konichezky et al.
Chest, 104(3), 971-973 (1993-09-01)
Thionyl-chloride (TCl) is used in the manufacture of lithium batteries, producing SO2 and HCl fumes on contact with water. We report two cases of accidental TCl exposure resulting in lung injury that may vary from a relatively mild and reversible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service