Skip to Content
Merck
All Photos(1)

Documents

30-1840

Sigma-Aldrich

Thionyl chloride

SAJ special grade, ≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SOCl2
CAS Number:
Molecular Weight:
118.97
Beilstein:
1209273
MDL number:
UNSPSC Code:
12352307
PubChem Substance ID:

grade

SAJ special grade

vapor pressure

97 mmHg ( 20 °C)

Assay

≥95.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.518 (lit.)

bp

79 °C (lit.)

mp

−105 °C (lit.)

density

1.631 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

ClS(Cl)=O

InChI

1S/Cl2OS/c1-4(2)3

InChI key

FYSNRJHAOHDILO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gwenaël Chamoulaud et al.
Langmuir : the ACS journal of surfaces and colloids, 20(12), 4989-4995 (2005-06-30)
This paper describes a novel approach for the surface modification of a cation-exchange membrane, bearing sulfonate groups, by a cationic layer. The modification procedure involved the chlorosulfonation of the sulfonate groups of the base membrane with thionyl chloride, followed by
Hend N Hafez et al.
Bioorganic & medicinal chemistry letters, 19(15), 4143-4147 (2009-06-23)
Chlorosulfonation of 3-methyl[1,2,4]triazolo[4,3-a]pyrimidine with chlorosulfonic acid in the presence of thionyl chloride was studied. When triazolo[4,3-a]pyrimidines are used as substrates, the substitution occurs at C-6. Also the reactivity of the hydrazides (7) towards aldehydes, thioglycolic acid and amines were studies.
Alexei Chichinin et al.
Physical chemistry chemical physics : PCCP, 7(2), 301-309 (2005-01-21)
New theoretical and experimental results for the ultraviolet photodissociation dynamics of thionyl chloride (SOCl2) are presented and combined with existing data from a variety of sources in order to provide a unified view of the photodissociation dynamics of SOC12. Time-dependent
Feng Xu et al.
The Journal of organic chemistry, 73(1), 312-315 (2007-12-07)
A simple, one-pot preparation of cyclic amines via efficient chlorination of amino alcohols with use of SOCl(2) has been developed. This approach obviates the need for the classical N-protection/O-activation/cyclization/deprotection sequence commonly employed for this type of transformation. The reaction pathways
S Aksoy et al.
Journal of biotechnology, 60(1-2), 37-46 (1998-05-08)
Poly(methyl methacrylate-acrylic acid) microspheres were prepared and the acid groups were activated by using either carbodiimide (CDI) or thionyl chloride (SOCl2). alpha-Amylase was covalently bound on these activated microspheres. The properties of the immobilized enzyme were investigated and compared with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service