Skip to Content
Merck
All Photos(1)

Key Documents

30-1650

Sigma-Aldrich

2,2′-Thiodiethanol

≥99.0%, suitable for amino acid analysis

Synonym(s):

2,2′-Thiobis(ethanol), Bis(2-hydroxyethyl) sulfide, Thiodiethylene glycol, Thiodiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S(CH2CH2OH)2
CAS Number:
Molecular Weight:
122.19
Beilstein:
1236325
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

Assay

≥99.0%

form

liquid

availability

available only in Japan

refractive index

n20/D 1.5215 (lit.)

bp

164-166 °C/20 mmHg (lit.)

mp

−16 °C (lit.)

density

1.221 g/mL at 25 °C (lit.)

suitability

suitable for amino acid analysis

SMILES string

OCCSCCO

InChI

1S/C4H10O2S/c5-1-3-7-4-2-6/h5-6H,1-4H2

InChI key

YODZTKMDCQEPHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

320.0 °F

Flash Point(C)

160 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Karvaly et al.
Journal of applied toxicology : JAT, 28(1), 21-26 (2007-04-13)
Subcutaneous microdialysis was employed for monitoring thiodiglycol (2,2'-thiodiethanol, TDG) levels with the aim of characterizing the transdermal penetration of topically applied liquid sulfur mustard (2,2'-dichlorodiethyl sulfide, SM) in rats. TDG levels, evaluated in 20 min dialysates collected over a 6
Karolin K Kroening et al.
Analytical and bioanalytical chemistry, 393(8), 1949-1956 (2009-02-14)
Sulfur mustard (HD), bis(2-chloroethyl)sulfide, is one of a class of mustard agents which are chemical warfare agents. The main chemical warfare hydrolysis degradation products of sulfur mustards are: thiodiglycol, bis(2-hydroxyethylthio)methane, 1,2-bis(2-hydroxyethylthio)ethane, 1,3-bis(2-hydroxyethylthio)propane, and 1,4-bis(2-hydroxyethylthio)butane. The aim of this study is
Leila El Bassi et al.
Journal of hazardous materials, 167(1-3), 124-127 (2009-01-31)
Microbial degradation of thiodiglycol (bis(2-hydroxyethyl)sulfide, TDG) with petroleum-desulfurizing soil bacteria was examined. Among the bacteria tested, several strains belonging to the genera Rhodococcus and Gordonia grew on TDG as the sole sulfur source. The selected strain Rhodococcus sp. strain T09
Hong Li et al.
Biodegradation, 24(1), 125-135 (2012-07-04)
Thiodiglycol (TDG) is both the precursor for chemical synthesis of mustard gas and the product of mustard gas hydrolysis. TDG can also react with intermediates of mustard gas degradation to form more toxic and/or persistent aggregates, or reverse the pathway
A A Brimfield et al.
Toxicology and applied pharmacology, 213(3), 207-215 (2006-01-19)
Thiodiglycol (2,2'-bis-hydroxyethylsulfide, TDG), the hydrolysis product of the chemical warfare agent sulfur mustard, has been implicated in the toxicity of sulfur mustard through the inhibition of protein phosphatases in mouse liver cytosol. The absence of any inhibitory activity when TDG

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service