Skip to Content
Merck
All Photos(1)

Documents

28-3400

Sigma-Aldrich

Sodium molybdate dihydrate

JIS special grade, ≥99.0%

Synonym(s):

Molybdic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2MoO4 · 2H2O
CAS Number:
Molecular Weight:
241.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: molybdenum

availability

available only in Japan

mp

100 °C (dec.) (lit.)

SMILES string

O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

InChI

1S/Mo.2Na.2H2O.4O/h;;;2*1H2;;;;/q;2*+1;;;;;2*-1

InChI key

FDEIWTXVNPKYDL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Padmarajaiah Nagaraja et al.
Journal of AOAC International, 85(4), 869-874 (2002-08-16)
A sensitive, rapid, and simple spectrophotometric method is described for the determination of sulfa drugs. The method is based on the formation of a red-colored product by the diazotization of sulfonamides such as sulfathiazole (SFT), sulfadiazine (SFD), sulfacetamide (SFA), sulfamethoxazole
Saraswathi Panneerselvam et al.
Chemico-biological interactions, 145(2), 159-163 (2003-04-11)
Diabetes mellitus is a disease, which virtually affects all the systems in the body including the immune system. Bacterial infections are important causes of increased morbidity and mortality in diabetic patients. In the present study, we assessed the effect of
S W Ball et al.
Canadian journal of physiology and pharmacology, 80(3), 205-209 (2002-05-07)
A focus of current diabetes research is the development of insulinomimetic compounds for oral treatment of diabetes and its associated cardiac complications. Screening compounds for their potential insulinomimetic effects usually involves the use of radioactive isotopes. The focus of this
Akira Sakakura et al.
Chemical communications (Cambridge, England), (30)(30), 3561-3563 (2008-07-26)
Efficient total syntheses of fluvibactin and vibriobactin have been achieved via molybdenum(VI) oxide-catalyzed dehydrative cyclization, Sb(OEt)(3)-catalyzed ester-amide transformation, and WSCI and HOAt-promoted dehydrative amide formation.
Laurent Caron et al.
Journal of colloid and interface science, 282(2), 478-485 (2004-12-14)
Chemically generated singlet oxygen (1O2, 1Deltag) is able to oxidize a great deal of hydrophobic substrates from molybdate-catalyzed hydrogen peroxide decomposition, provided a suitable reaction medium such as a microemulsion system is used. However, high substrate concentrations or poorly reactive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service