Skip to Content
Merck
All Photos(1)

Key Documents

28-2070

Sigma-Aldrich

Sodium bromide

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.0%
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

1 mmHg ( 806 °C)

Assay

≥99.0%

form

solid

availability

available only in Japan

pH

5.4 (20 °C, 50 g/L)

mp

755 °C (lit.)

storage temp.

15-25°C

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian Holley et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 84(5 Pt 2), 056110-056110 (2011-12-21)
Excitation waves on a subexcitable Belousov-Zhabotinsky (BZ) substrate can be manipulated by chemical variations in the substrate and by interactions with other waves. Symbolic assignment and interpretation of wave dynamics can be used to perform logical and arithmetic computations. We
David G Levitt et al.
Journal of applied physiology (Bethesda, Md. : 1985), 109(3), 786-795 (2010-06-19)
Measurement of body composition changes following bariatric surgery is complicated because of the difficulty of measuring body fat in highly obese individuals that have increased photon absorption and are too large for the standard dual-energy X-ray absorptiometry (DXA) table. We
Kajari Maiti et al.
The journal of physical chemistry. B, 114(22), 7499-7508 (2010-05-19)
Bolaforms B(1), B(2), and B(3) of the formulas, Br(-)Me(3)N(+)(CH(2))(10)N(+)Me(3)Br(-), Br(-)Me(3)N(+)(CH(2))(10)OH, and Br(-)Me(3)N(+)(CH(2))(10)COO(-)Na(+), respectively, were synthesized, and their properties in the bulk as well as at the air/aqueous NaBr (10 mM) solution interface have been studied. Their interactions with sodium dodecyl
Jun Huang et al.
Carbohydrate polymers, 91(1), 191-197 (2012-10-10)
TEMPO (2,2,6,6-tetramethylpiperidine-1-oxyl radical)-mediated 6-carboxy β-chitin derivatives (T-chitin) with different carboxylate content were successfully synthesized by controlling the addition level of NaClO as the primary oxidant. The structural and biochemical properties of the derivatives were investigated. The carboxylate contents of the
Carlos Barata et al.
Environmental science & technology, 46(17), 9663-9672 (2012-07-26)
When considering joint toxic apical effects at higher levels of biological organization, such as the growth of populations, the so-called pharmacological mode of action that relies on toxicological mechanistic effects on molecular target sites may not be relevant. Such effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service