Skip to Content
Merck
All Photos(1)

Documents

28-1580

Sigma-Aldrich

Sodium acetate trihydrate

≥99.0%, suitable for amino acid analysis

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.0%

form

solid

autoignition temp.

1112 °F

availability

available only in Japan

pH

8.5-10 (25 °C, 408 g/L)

suitability

suitable for amino acid analysis

storage temp.

15-25°C

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace
Do Gyun Lee et al.
Water research, 46(13), 4226-4234 (2012-06-08)
Triclosan, a synthetic antimicrobial agent, has been considered as an emerging environmental contaminant. Here we reported a triclosan-degrading wastewater bacterial isolate, Sphingopyxis strain KCY1, capable of dechlorinating triclosan with a stoichiometric release of chloride. The stain can degrade diphenyl ether
Kit Frederiksen et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 91, 9-15 (2015-01-18)
Polymeric film-forming systems (FFSs) are potential drug delivery systems for topical application to the skin. The FFSs form thin and transparent polymeric films in situ upon solvent evaporation. Their application convenience and cosmetic attributes, superior to conventional semi-solids, may offer
Rishiram Ramanan et al.
FEBS letters, 587(4), 370-377 (2013-01-15)
Phenotypic and genotypic changes in Chlamydomonas reinhardtii BafJ5, a starchless mutant, with respect to lipid metabolism was studied in different trophic states under nitrogen (N) sufficient and limited conditions. Interestingly, cellular lipid content increased linearly with input acetate concentration with
Horng-Ruey Chua et al.
Journal of critical care, 27(2), 138-145 (2012-03-24)
The purpose of the study was to determine the effects of Plasma-Lyte 148 (PL) vs 0.9% saline (NS) fluid resuscitation in diabetic ketoacidosis (DKA). A multicenter retrospective analysis of adults admitted for DKA to the intensive care unit, who received

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service