Skip to Content
Merck
All Photos(1)

Documents

24-6250

Sigma-Aldrich

Propyl 4-hydroxybenzoate

SAJ first grade, ≥98.0%

Synonym(s):

4-Hydroxybenzoic acid propyl ester, Propyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH2CH2CH3
CAS Number:
Molecular Weight:
180.20
Beilstein:
1103245
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

95-98 °C (lit.)

SMILES string

CCCOC(=O)c1ccc(O)cc1

InChI

1S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11H,2,7H2,1H3

InChI key

QELSKZZBTMNZEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

356.0 °F

Flash Point(C)

180 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dennis G Maki et al.
Critical care medicine, 39(4), 613-620 (2011-01-05)
Catheter-related bloodstream infection is the greatest threat to the safety of patients on hemodialysis. Catheter lock solutions containing heparin have been linked to an increased risk of hemorrhage and thrombocytopenia. To ascertain the safety and efficacy for prevention of catheter-related
C L Barberis et al.
Journal of food protection, 73(8), 1493-1501 (2010-09-08)
Each year, a significant portion of the peanuts produced cannot be marketed because of fungal disease at the postharvest stage and mycotoxin contamination. Antioxidants could be used as an alternative to fungicides to control ochratoxigenic fungi in peanuts during storage.
Aeshna Amin et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 75(2), 206-212 (2010-03-09)
p-Hydroxybenzoic acid esters (parabens) are commonly used antimicrobial preservatives in pharmaceutical formulations. Two microorganisms, isolated from non-sterile methyl paraben (MP) and propyl paraben (PP) solutions, were found to degrade the respective parabens. Identification by 16S rRNA partial gene sequencing revealed
Miguel González-Doncel et al.
Environmental pollution (Barking, Essex : 1987), 184, 360-369 (2013-10-08)
Here we proposed a battery of non-invasive biomarkers and a histological survey to examine physiological/anatomical features in embryos, eleutheroembryos (13 days post-fertilization, dpf), and larvae (28-42 dpf) of medaka to investigate the effects of embryonic exposure to propylparaben (PrP). Concentrations <1000 μg
Yang Wang et al.
Analytica chimica acta, 673(2), 145-150 (2010-07-06)
A selective, sensitive, rapid and reliable method based on molecularly imprinted polymers (MIPs) with dual templates to determine total content of parabens in cosmetics was developed. With methylparaben (MP) and propylparaben (PP) as dual-templates, methacrylic acid (MAA) as a functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service