Skip to Content
Merck
All Photos(1)

Key Documents

24-5630

Sigma-Aldrich

Potassium sulfide

Potassium sulfide

CP

Synonym(s):

Dipotassium monosulfide, Dipotassium sulfide, Potassium monosulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S
Molecular Weight:
110.26
UNSPSC Code:
12352300
grade:
CP
form:
solid
Pricing and availability is not currently available.

grade

CP

form

solid

availability

available only in Japan

storage temp.

15-25°C

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 福岡
    • Review 1
    • Votes 4
    1 out of 5 stars.

    K2Sはほぼ含まれていません

    標記の通りK2Sだと思い購入しましたが、他硫化物が実情です。
    カスタマーサービス連絡すると、純度の表記がないので嘘ではない、とのことでした。
    注意喚起の意味も込めて、ここにレビューを残しておきます。

    Translate with Google

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for your review. We are sorry to hear that your experience did not match your product expectations. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service