Skip to Content
Merck
All Photos(1)

Key Documents

21-1260

Sigma-Aldrich

Nickel(II) nitrate hexahydrate

SAJ special grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
290.79
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥98.0%
grade:
SAJ special grade
form:
crystalline

grade

SAJ special grade

Assay

≥98.0%

form

crystalline

reaction suitability

reagent type: catalyst
core: nickel

availability

available only in Japan

mp

56 °C (lit.)

density

2.05 g/mL at 25 °C (lit.)

SMILES string

O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Ni.6H2O/c2*2-1(3)4;;;;;;;/h;;;6*1H2/q2*-1;+2;;;;;;

InChI key

AOPCKOPZYFFEDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Débora R Lima et al.
ACS applied materials & interfaces, 9(45), 39830-39838 (2017-10-24)
CoFe
Sheng-Mu You et al.
Nanomaterials (Basel, Switzerland), 10(9) (2020-09-02)
Photoelectrochemical (PEC) water splitting is a promising strategy to improve the efficiency of oxygen evolution reactions (OERs). However, the efficient adsorption of visible light as well as long-term stability of light-harvesting electrocatalysis is the crucial issue in PEC cells. Metal-organic
H Tanojo et al.
Acta dermato-venereologica. Supplementum, (212)(212), 19-23 (2002-01-23)
Allergic contact dermatitis due to nickel salts is common. It is therefore important to measure the permeation of these salts through the stratum corneum (SC), the primary rate-limiting domain in skin. An advanced diffusion system and analytical techniques now enable
J-C Amiard et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 147(3), 378-385 (2008-02-16)
In mussels impacted by the V- and Ni-rich oil dispersed in the environment after the < Erika > wreck, a significant and positive correlation was observed between metallothionein (MT) and V concentrations whereas no correlation was observed between MT and
Jun Li et al.
Journal of colloid and interface science, 332(1), 231-236 (2009-01-16)
In this paper, we report the successful synthesis of Ni(12)P(5) hollow spheres via a facile hydrothermal route, employing white phosphorus (WP) and nickel nitrate as the reactants in the presence of hexamethylenetetramine (HMT) and polyethylene glycol 10000 (PEG-10000). The phase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service