Skip to Content
Merck
All Photos(1)

Key Documents

19-2340

Sigma-Aldrich

Methyl acetate

JIS special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH3
CAS Number:
Molecular Weight:
74.08
Beilstein:
1736662
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
grade:
JIS special grade
bp:
57-58 °C (lit.)
vapor pressure:
165 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

JIS special grade

vapor density

2.55 (vs air)

vapor pressure

165 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

936 °F

expl. lim.

16 %

availability

available only in Japan

refractive index

n20/D 1.361 (lit.)

bp

57-58 °C (lit.)

mp

−98 °C (lit.)

density

0.934 g/mL at 25 °C

SMILES string

COC(C)=O

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Yang et al.
The journal of physical chemistry. A, 112(28), 6364-6372 (2008-06-21)
The mechanisms and the kinetics of the OH (OD) radicals with methyl acetate CH3C(O)OCH3 are investigated theoretically. The dual-level direct dynamics method is employed in the calculation of the rate constants. The optimized geometries and frequencies and the gradients of
Nicolas Salem et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 13(1), 140-151 (2010-04-20)
Studies have established the value of [(methyl)1-(11)C]-acetate ([(11)C]Act) combined with 2-deoxy-2[(18)F]fluoro-D-glucose (FDG) for detecting hepatocellular carcinoma (HCC) using positron emission tomography (PET). In this study, the metabolic fate of [(11)C]Act in HCC was characterized. Experiments with acetic acid [1-(14)C] sodium
Benjamin Bechem et al.
The Journal of organic chemistry, 75(5), 1795-1798 (2010-02-06)
5-Substituted-2-furan methanols 1a-c are subject to enantioselective carbonyl allylation, crotylation and tert-prenylation upon exposure to allyl acetate, alpha-methyl allyl acetate, or 1,1-dimethylallene in the presence of an ortho-cyclometalated iridium catalyst modified by (R)-Cl,MeO-BIPHEP, (R)-C3-TUNEPHOS, and (R)-C3-SEGPHOS, respectively. In the presence
Saman Mohammadi et al.
PloS one, 9(9), e106653-e106653 (2014-09-11)
In this study, we compared, for the first time, the release of a 432 kDa prostaglandin F2a analogue drug, Latanoprost, from commercially available contact lenses using in vitro models with corneal epithelial cells. Conventional polyHEMA-based and silicone hydrogel soft contact
Franc Avbelj et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(9), 3137-3141 (2009-02-10)
Recent calorimetric measurements of the solvation enthalpies of some dipeptide analogs confirm our earlier prediction that the principle of group additivity is not valid for the interaction of the peptide group with water. We examine the consequences for understanding the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service