Skip to Content
Merck
All Photos(1)

Key Documents

19-0540

Sigma-Aldrich

Malachite Green oxalate

SAJ first grade, ≥80.0%

Synonym(s):

Basic Green 4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H54N4O12
CAS Number:
Molecular Weight:
927.00
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥80.0%

form

solid

availability

available only in Japan

mp

164 °C (dec.) (lit.)

SMILES string

OC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)c1ccc(cc1)C(\c2ccccc2)=C3\C=CC(\C=C3)=[N+](\C)C.CN(C)c4ccc(cc4)C(\c5ccccc5)=C6\C=CC(\C=C6)=[N+](\C)C

InChI

1S/2C23H25N2.3C2H2O4/c2*1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4;3*3-1(4)2(5)6/h2*5-17H,1-4H3;3*(H,3,4)(H,5,6)/q2*+1;;;/p-2

InChI key

CNYGFPPAGUCRIC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Sahraei et al.
Environmental monitoring and assessment, 185(7), 5817-5822 (2012-12-04)
A new spectrophotometric method is reported for the determination of nanomolar level of malachite green in surface water samples. The method is based on the catalytic effect of silver nanoparticles on the oxidation of malachite green by hexacyanoferrate (III) in
P Manoj Kumar Reddy et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(5), 1097-1104 (2013-02-19)
Oxidative decomposition of aqueous organic pollutant malachite green (MG) was studied in a dielectric barrier discharge reactor operated under ambient conditions. Total organic carbon content analysis confirmed the mineralization of the pollutant leading to the formation of carbon dioxide, which
Xiaofang Jia et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(42), 13494-13500 (2012-09-05)
This work describes a novel strategy for the highly sensitive and selective detection of cysteine (Cys) and glutathione (GSH) based on the Hg(2+)-AGRO100-malachite green (MG) complex system. The dye MG, which has a very low quantum yield in aqueous solution
Jia Zeng et al.
Biophysical journal, 104(1), 139-145 (2013-01-22)
It is shown that the nonlinear optical phenomenon known as second-harmonic generation can be used for label-free, time-resolved study of the transport of molecules through living cell membranes. The adsorption and transport of a 300-Da molecular-mass hydrophobic ion at the
Yang Wu et al.
Cytometry. Part A : the journal of the International Society for Analytical Cytology, 83(2), 220-226 (2013-01-11)
Monitoring the trafficking of multiple proteins simultaneously in live cells is of great interest because many receptor proteins are found to function together with others in the same cell. However, existing fluorescent labeling techniques have restricted the mechanistic study of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service