Skip to Content
Merck
All Photos(1)

Key Documents

15-0600

Sigma-Aldrich

Iodine

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2
CAS Number:
Molecular Weight:
253.81
Beilstein:
3587194
EC Number:
MDL number:
UNSPSC Code:
12141916
PubChem Substance ID:

grade

SAJ first grade

vapor density

9 (vs air)

vapor pressure

0.31 mmHg ( 25 °C)
1 mmHg ( 38.7 °C)

Assay

≥99.0%

availability

available only in Japan

resistivity

1.3E15 μΩ-cm

bp

184 °C (lit.)

mp

113 °C (lit.)

SMILES string

II

InChI

1S/I2/c1-2

InChI key

PNDPGZBMCMUPRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Respiratory system, Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eike I Piechowiak et al.
Radiology, 275(3), 692-697 (2015-02-06)
To determine the effect of the use of iodinated contrast agents on the formation of DNA double-strand breaks during chest computed tomography (CT). This study was approved by the institutional review board, and written informed consent was obtained from all
Tim I M Korevaar et al.
The Journal of clinical endocrinology and metabolism, 98(11), 4382-4390 (2013-09-17)
Premature delivery is an important risk factor for child mortality and psychiatric, metabolic, and cardiovascular disease later in life. In the majority of cases, the cause of prematurity cannot be identified. Currently, it remains controversial whether abnormal maternal thyroid function
Zulfiqar A Bhutta et al.
Lancet (London, England), 382(9890), 452-477 (2013-06-12)
Maternal undernutrition contributes to 800,000 neonatal deaths annually through small for gestational age births; stunting, wasting, and micronutrient deficiencies are estimated to underlie nearly 3·1 million child deaths annually. Progress has been made with many interventions implemented at scale and
Moritz Tebbe et al.
Faraday discussions, 181, 243-260 (2015-05-08)
We demonstrate the large-scale organisation of anisotropic nanoparticles into linear assemblies displaying optical anisotropy on macroscopic areas. Monodisperse gold nanorods with a hydrophilic protein shell are arranged by dip-coating on wrinkled surfaces and subsequently transferred to indium tin oxide (ITO)
Astri Handayani et al.
Investigative radiology, 50(4), 275-282 (2014-11-25)
The aim of this study was to assess the intermodel agreement of different magnetic resonance myocardial perfusion models and evaluate their correspondence to stenosis diameter. In total, 260 myocardial segments were analyzed from rest and adenosine stress first-pass myocardial perfusion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service