Skip to Content
Merck
All Photos(1)

Key Documents

13-1580

Sigma-Aldrich

Hydriodic acid

JIS special grade, 55.0-58.0%

Synonym(s):

Hydriotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HI
CAS Number:
Molecular Weight:
127.91
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
concentration:
55.0-58.0%
grade:
JIS special grade

grade

JIS special grade

form

liquid

availability

available only in Japan

concentration

55.0-58.0%

bp

127 °C (lit.)

SMILES string

I

InChI

1S/HI/h1H

InChI key

XMBWDFGMSWQBCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazumasa Wakamatsu et al.
Pigment cell & melanoma research, 22(4), 474-486 (2009-06-06)
Pheomelanogenesis is a complex pathway that starts with the oxidation of tyrosine (or DOPA, 3,4-dihydroxyphenylalanine) by tyrosinase in the presence of cysteine, which results in the production of 5-S-cysteinyldopa and its isomers. Beyond that step, relatively little has been clarified
Relationships between circulating and intracellular thyroid hormones: physiological and clinical implications.
P R Larsen et al.
Endocrine reviews, 2(1), 87-102 (1981-01-01)
Marina M Demidova et al.
The American journal of cardiology, 115(4), 417-422 (2015-01-01)
Ventricular fibrillation (VF) during reperfusion (rVF) in ST-segment elevation myocardial infarction (STEMI) is an infrequent but serious event that complicates coronary interventions. The aim of this study was to analyze clinical predictors of rVF in an unselected population of patients
Xin Yu Chin et al.
Nature communications, 6, 7383-7383 (2015-06-26)
Despite the widespread use of solution-processable hybrid organic-inorganic perovskites in photovoltaic and light-emitting applications, determination of their intrinsic charge transport parameters has been elusive due to the variability of film preparation and history-dependent device performance. Here we show that screening
Daniel J Weisenberger et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 24(3), 512-519 (2015-01-15)
The CpG island methylator phenotype (CIMP) represents a subset of colorectal cancers characterized by widespread aberrant DNA hypermethylation at select CpG islands. The risk factors and environmental exposures contributing to etiologic heterogeneity between CIMP and non-CIMP tumors are not known.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service