Skip to Content
Merck
All Photos(1)

Key Documents

13-1130

Sigma-Aldrich

Histamine dihydrochloride

SAJ special grade, ≥98.0%

Synonym(s):

2-(4-Imidazolyl)ethylamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3 · 2HCl
CAS Number:
Molecular Weight:
184.07
Beilstein:
3624116
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
SAJ special grade
Assay:
≥98.0%

grade

SAJ special grade

Assay

≥98.0%

form

solid

availability

available only in Japan

mp

249-252 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].NCCc1c[nH]cn1

InChI

1S/C5H9N3.2ClH/c6-2-1-5-3-7-4-8-5;;/h3-4H,1-2,6H2,(H,7,8);2*1H

InChI key

PPZMYIBUHIPZOS-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Bessho et al.
Neuroscience letters, 578, 106-110 (2014-07-06)
In mammals, it is established that histamine is a neurotransmitter and/or neuromodulator in the central nervous system. It is produced by the enzyme histidine decarboxylase (HDC) in the tuberomammillary nucleus of the posterior hypothalamus. However, HDC as well as histaminergic
Chayon Goswami et al.
PloS one, 10(6), e0131554-e0131554 (2015-06-27)
Motilin and ghrelin constitute a peptide family, and these hormones are important for the regulation of gastrointestinal motility. In this study, we examined the effect of motilin and ghrelin on gastric acid secretion in anesthetized suncus (house musk shrew, Suncus
Kevin B Freeman et al.
Psychopharmacology, 231(12), 2395-2403 (2014-01-09)
Recent research has demonstrated that the drug, histamine, can function as a punisher of cocaine self-administration. However, little is known about how drug punishers affect the maximum reinforcing effectiveness of drugs as reinforcers. The goal of the present study was
Guadalupe-Elide Morales-Figueroa et al.
ACS chemical neuroscience, 5(8), 637-645 (2014-06-03)
High levels of histamine H3 receptors (H3Rs) are found in the globus pallidus (GP), a neuronal nucleus in the basal ganglia involved in the control of motor behavior. By using rat GP isolated nerve terminals (synaptosomes), we studied whether H3R
Nico A Jansen et al.
Journal of neurophysiology, 113(1), 58-70 (2014-10-10)
We tested the possibility that the trigeminoparabrachial tract (VcPbT), a projection thought to be importantly involved in nociception, might also contribute to sensation of itch. In anesthetized rats, 47 antidromically identified VcPbT neurons with receptive fields involving the cheek were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service