Skip to Content
Merck
All Photos(1)

Key Documents

09-1833

Sigma-Aldrich

Diethyl ether

≥99.5% (GC), for residue analysis, JIS 5000

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

Product Name

Diethyl ether, for residue analysis, JIS 5000

grade

for residue analysis

Agency

JIS 5000

vapor density

2.6 (vs air)

vapor pressure

28.5 psi

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

320 °F

expl. lim.

36.5 %

availability

available only in Japan

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

suitability

suitable for residue analysis (suitable for 5000 per JIS)
suitable for residue analysis

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-49.0 °F

Flash Point(C)

-45 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan E Camacho Londoño et al.
European heart journal, 36(33), 2257-2266 (2015-06-13)
Pathological cardiac hypertrophy is a major predictor for the development of cardiac diseases. It is associated with chronic neurohumoral stimulation and with altered cardiac Ca(2+) signalling in cardiomyocytes. TRPC proteins form agonist-induced cation channels, but their functional role for Ca(2+)
Alessandra Forni et al.
Journal of molecular graphics & modelling, 38, 31-39 (2012-10-23)
The solvent effect on the I⋯O halogen bonding in complexes of iodobenzene derivatives with formaldehyde has been investigated by systematically varying the substituents on the iodobenzene ring. Calculations have been performed at MP2 and DFT levels of theory, using the
Faysal Benaskar et al.
ChemSusChem, 6(2), 353-366 (2012-11-30)
A μ(2)-process in the Ullmann-type C-O coupling of potassium phenolate and 4-chloropyridine was successfully performed in a combined microwave (MW) and microflow process. Selective MW absorption in a micro-fixed-bed reactor (μ-FBR) by using a supported Cu nanocatalyst resulted in an
Ether-directed ortho-C-H olefination with a palladium(II)/monoprotected amino acid catalyst.
Gang Li et al.
Angewandte Chemie (International ed. in English), 52(4), 1245-1247 (2012-12-15)
Tatsuya Yoshino et al.
Organic letters, 14(16), 4290-4292 (2012-08-09)
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service