Skip to Content
Merck
All Photos(1)

Key Documents

09-1260

Sigma-Aldrich

2-Chloroethanol

SAJ special grade, ≥99.0%

Synonym(s):

Ethylene chlorohydrin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2OH
CAS Number:
Molecular Weight:
80.51
Beilstein:
878139
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

SAJ special grade

vapor density

2.78 (vs air)

vapor pressure

5 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

797 °F

expl. lim.

16 %

availability

available only in Japan

refractive index

n20/D 1.441 (lit.)

bp

129 °C (lit.)

mp

−89 °C (lit.)

density

1.201 g/mL at 25 °C (lit.)

SMILES string

OCCCl

InChI

1S/C2H5ClO/c3-1-2-4/h4H,1-2H2

InChI key

SZIFAVKTNFCBPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Bednar et al.
Journal of hazardous materials, 168(2-3), 1041-1046 (2009-04-07)
Active treatment of BCEE (bis-2-chloroethyl ether) is being currently performed in the on-site Cohansey Aquifer at the Lipari Superfund Site. Remediation of BCEE in the underlying Kirkwood aquifer is being considered, necessitating investigations of BCEE geochemistry in aquifer material from
Nikhil K Parelkar et al.
The Journal of pharmacology and experimental therapeutics, 332(3), 803-810 (2009-12-04)
Trichloroacetaldehyde monohydrate [chloral hydrate (CH)] is a sedative/hypnotic that increases cerebral blood flow (CBF), and its active metabolite 2,2,2-trichloroethanol (TCE) is an agonist for the nonclassical two-pore domain K(+) (K(2P)) channels TREK-1 and TRAAK. We sought to determine whether TCE
Sarah B Appel et al.
The Journal of pharmacology and experimental therapeutics, 318(3), 1137-1145 (2006-06-03)
The dopaminergic neurons of the ventral tegmental area (DA VTA neurons) have been implicated in the reinforcing properties of drugs of abuse, including ethanol (ethyl alcohol). Ethanol increases the spontaneous firing frequency of DA VTA neurons in vitro, in both
J L Merdink et al.
Toxicology, 245(1-2), 130-140 (2008-02-05)
Chloral hydrate (CH) is a short-lived intermediate in the metabolism of trichloroethylene (TRI). TRI, CH, and two common metabolites, trichloroacetic acid (TCA) and dichloroacetic acid (DCA) have been shown to be hepatocarcinogenic in mice. To better understand the pharmacokinetics of
Haijun Yang et al.
Toxicology in vitro : an international journal published in association with BIBRA, 25(8), 1638-1643 (2011-07-12)
Occupational trichloroethylene (TCE) exposure could induce generalized skin hypersensitivity reactions complicated with severe liver dysfunctions. Active extracellular matrix degradation and remodeling are involved in the skin hypersensitivity reaction induced by chemical exposure. In the present study, we have compared the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service