Journal of agricultural and food chemistry, 58(11), 6556-6561 (2010-05-15)
A simple colorimetric method for the differentiation of indoleacetic acid (IAA) and indolebutyric acid (IBA) in plant samples is described. The color change is based upon the reaction between the auxins and p-(dimethylamino)benzaldehyde (PDAB, Ehrlich reagent) following the electrophilic substitution
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 54A(6), 799-819 (1998-07-17)
Results of a normal coordinate analysis based on infrared and Raman spectra of six isotopic forms of 4-(dimethylamino)benzaldehyde are reported (DABA, DABA-CDO, DABA-13CDO, DABA-13CHO, DABA-CHO18O, and DABA-3,5-D). Cs point group symmetry has been applied and all motions except the internal
Spectrophotometric determination of certain cephalosporins through oxidation with cerium(IV) and 1-chlorobenzotriazole.
M M Ayad et al.
Journal of pharmaceutical and biomedical analysis, 20(3), 557-564 (2000-03-04)
The Journal of biological chemistry, 275(45), 34873-34880 (2000-08-24)
FlaA1 is a small soluble protein of unknown function in Helicobacter pylori. It has homologues that are essential for the virulence of numerous medically relevant bacteria. FlaA1 was overexpressed as a histidine-tagged protein and purified to homogeneity by nickel chelation
Biochimica et biophysica acta, 1502(3), 398-404 (2000-11-09)
In the present work, pepsin digests of tail tendons from streptozotocin-diabetic rats were found to contain material that reacted rapidly at room temperature with p-dimethylaminobenzaldehyde (Ehrlich's reagent) to give an adduct with an absorbance spectrum characteristic of the Ehrlich chromogen
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.