Skip to Content
Merck
All Photos(1)

Key Documents

07-2960

Sigma-Aldrich

Diethylene glycol butyl ether

SAJ special grade, ≥99.0%

Synonym(s):

2-(2-Butoxyethoxy)ethanol, BDG, Butyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
162.23
Beilstein:
1739225
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
grade:
SAJ special grade
application(s):
hematology
bp:
231 °C (lit.)
vapor pressure:
30 mmHg ( 130 °C)
Pricing and availability is not currently available.

grade

SAJ special grade

vapor density

5.6 (vs air)

vapor pressure

30 mmHg ( 130 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

400 °F

expl. lim.

6.2 %

availability

available only in Japan

refractive index

n20/D 1.432 (lit.)

pH

7

bp

231 °C (lit.)

mp

−68 °C (lit.)

density

0.967 g/mL at 25 °C (lit.)

application(s)

hematology

SMILES string

CCCCOCCOCCO

InChI

1S/C8H18O3/c1-2-3-5-10-7-8-11-6-4-9/h9H,2-8H2,1H3

InChI key

OAYXUHPQHDHDDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeomin Yoon et al.
Journal of membrane science, 261(1-2), 76-86 (2005-09-02)
Rejection characteristics of organic and inorganic compounds were examined for six reverse osmosis (RO) membranes and two nanofiltration (NF) membranes that are commercially available. A batch stirred-cell was employed to determine the membrane flux and the solute rejection for solutions
Thomas Göen et al.
Toxicology letters, 134(1-3), 295-300 (2002-08-23)
The aim of the study was to investigate the suitability of butoxyethoxyacetic acid (BEAA) as a biomarker of exposure to water-based cleaning agents containing diethylene glycol mono butyl ether (DEGBE). The study was performed in two printing plants where water-based
Christopher R Gibson et al.
Journal of the American Society for Mass Spectrometry, 14(11), 1247-1249 (2003-11-05)
Tandem liquid chromatography-mass spectrometry coupled to online radioactive material detection (LC/RAM/MS/MS) is a technique that is used routinely for in vivo and in vitro drug metabolism studies and allows for a simultaneous correlation between radiochemical peaks and mass spectral data.
M Ema et al.
Drug and chemical toxicology, 11(2), 97-111 (1988-06-01)
The teratogenicity of diethylene glycol mono-n-butyl ether (DEGMBE) was studied in Wistar rats. The pregnant rats were fed a diet containing DEGMBE from day 0 through day 20 of pregnancy. The dietary concentrations of DEGMBE were 0, 0.04, 0.2 and
Delayed and immediate hypersensitivity to carbitols.
T A Dawson et al.
Contact dermatitis, 21(1), 52-53 (1989-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service