Skip to Content
Merck
All Photos(1)

Key Documents

05-6080

Sigma-Aldrich

Copper(II) oxide

CP

Synonym(s):

Cupric oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

form

powder

reaction suitability

reagent type: catalyst
core: copper

availability

available only in Japan

SMILES string

[Cu]=O

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisha Zhou et al.
Nanoscale, 5(4), 1564-1569 (2013-01-18)
Stable Cu(2)O nanocrystals of around 3 nm were uniformly and densely grown on functionalized graphene sheets (FGS), which act as molecular templates instead of surfactants for controlled nucleation; the distribution density of nanocrystals can be easily controlled by FGS with
Hongwen Huang et al.
Nanoscale, 5(5), 1785-1788 (2013-01-31)
A facile, flexible and large-scale technique was proposed to prepare a CuO-CNT 3D-network composite with the aid of electrostatic interactions in aqueous solution. The composite greatly improves the electrochemical performance. At a rate of 0.1 C, the cycling discharge capacity
Wei Jian Foo et al.
Nanoscale, 5(2), 759-764 (2012-12-12)
Here we have demonstrated the preparation of high-quality, monodispersed and tunable phases of Cu nanoparticles. Structural and chemical composition studies depict the evolution of Cu-Cu(2)O-CuO nanoparticles at various process stages. The loading of Cu and Cu oxide nanoparticles on TiO(2)
Nicola Scotti et al.
Chemical communications (Cambridge, England), 49(19), 1957-1959 (2013-01-30)
The epoxidation of alkenes was carried out over a CuO/Al(2)O(3) catalyst using cumene as an oxygen carrier, through a one-pot reaction, giving high conversion and selectivity with different substrates. Trans-β-methylstyrene gave the corresponding epoxide in 95% yield after 3 h.
Qiao bao Zhang et al.
Nanotechnology, 24(6), 065602-065602 (2013-01-24)
Reversible superhydrophobic and superhydrophilic surfaces based on porous substrates covered with CuO nanowires are developed in this study. A facile thermal oxidation method is used to synthesize non-flaking bicrystalline CuO nanowires on porous copper substrates in static air. The effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service