Skip to Content
Merck
All Photos(1)

Key Documents

05-0015

Sigma-Aldrich

Cacodylic acid

SAJ first grade

Synonym(s):

Dimethylarsinic acid, Dimethylarsonic acid, Hydroxydimethylarsine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2As(O)OH
CAS Number:
Molecular Weight:
138.00
Beilstein:
1736965
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

form

solid

reaction suitability

reagent type: catalyst
core: arsenic

availability

available only in Japan

SMILES string

C[As](C)(O)=O

InChI

1S/C2H7AsO2/c1-3(2,4)5/h1-2H3,(H,4,5)

InChI key

OGGXGZAMXPVRFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiro Narukawa et al.
Journal of agricultural and food chemistry, 60(4), 1122-1127 (2012-01-10)
A monitoring test for arsenic species in white rice flour was developed and applied to flours made from 20 samples of polished rice collected from locations all over Japan. The arsenic species in white rice flour made from five samples
Kavitha Marapakala et al.
Biochemistry, 51(5), 944-951 (2012-01-20)
The enzyme As(III) S-adenosylmethionine methyltransferase (EC 2.1.1.137) (ArsM or AS3MT) is found in members of every kingdom, from bacteria to humans. In these enzymes, there are three conserved cysteine residues at positions 72, 174, and 224 in the CmArsM orthologue
Hua Naranmandura et al.
Analytical and bioanalytical chemistry, 405(6), 1885-1891 (2012-06-27)
Arsenic (+3 oxidation state) methyltransferase (As3mt) plays a central role in the enzymatically catalyzed conversion of inorganic arsenic into methylated metabolites. Most studies of the metabolism and disposition of arsenicals following exposure to inorganic arsenic focus on the formation and
Matthew A Davis et al.
Environmental health perspectives, 120(10), 1418-1424 (2012-09-26)
In adult populations, emerging evidence indicates that humans are exposed to arsenic by ingestion of contaminated foods such as rice, grains, and juice; yet little is known about arsenic exposure among children. Our goal was to determine whether rice consumption
Erik J Tokar et al.
Toxicology letters, 209(2), 179-185 (2012-01-11)
Inorganic arsenic, an early life carcinogen in humans and mice, can initiate lesions promotable by other agents in later life. The biomethylation product of arsenic, dimethylarsinic acid (DMA), is a multi-site tumor promoter. Thus, pregnant CD1 mice were given drinking

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service