Skip to Content
Merck
All Photos(1)

Key Documents

03-0550

Sigma-Aldrich

Barium sulfate

SAJ special grade

Synonym(s):

Baryte

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaSO4
CAS Number:
Molecular Weight:
233.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ special grade

form

powder

availability

available only in Japan

pH

7

SMILES string

[Ba++].[O-]S([O-])(=O)=O

InChI

1S/Ba.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

TZCXTZWJZNENPQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George X Huang et al.
Tissue engineering. Part A, 21(15-16), 2228-2240 (2015-04-30)
Polymeric scaffolds, which release growth factors in a temporally controlled manner, have successfully directed the differentiation of stem cells into monolithic tissues of a single lineage. However, engineering precise boundaries in multilineage functional tissues, such as the juxtaposed cartilaginous and
Craig Ungaro et al.
Optics express, 23(19), A1149-A1156 (2015-09-26)
This paper presents results on a highly efficient experimental solar thermophotovoltaic (STPV) system using simulated solar energy. An overall power conversion efficiency of 6.2% was recorded under solar simulation. This was matched with a thermodynamic model, and the losses within
Gregory S Cooper et al.
Cancer, 119(10), 1800-1807 (2013-02-26)
Professional society guidelines recommend follow-up colonoscopy for patients with resected colonic adenomas. However, adherence to guideline recommendations in routine clinical practice has not been well characterized. The authors used a population-based sample of Medicare beneficiaries to identify all patients aged
Nam P Nguyen et al.
PloS one, 8(3), e56290-e56290 (2013-03-19)
The study aims to assess the feasibility of Tomotherapy-based image-guided radiotherapy (IGRT) to reduce the aspiration risk in patients with non-laryngeal and non-hypopharyngeal cancer. A retrospective review of 48 patients undergoing radiation for non-laryngeal and non-hypopharyngeal head and neck cancers
Steve Halligan et al.
Lancet (London, England), 381(9873), 1185-1193 (2013-02-19)
Barium enema (BE) is widely available for diagnosis of colorectal cancer despite concerns about its accuracy and acceptability. Computed tomographic colonography (CTC) might be a more sensitive and acceptable alternative. We aimed to compare CTC and BE for diagnosis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service