Skip to Content
Merck
All Photos(1)

Key Documents

01-6430

Sigma-Aldrich

L-Arginine

SAJ special grade, ≥98.0%

Synonym(s):

(S)-2-Amino-5-guanidinopentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
174.20
Beilstein:
1725413
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

SAJ special grade

Assay

≥98.0%

availability

available only in Japan

mp

222 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1

InChI key

ODKSFYDXXFIFQN-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Substrate of nitric oxide synthase, which is converted to citrulline and nitric oxide (NO). Induces insulin release by a nitric oxide-dependent mechanism.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palaniraja Thandapani et al.
Molecular cell, 50(5), 613-623 (2013-06-12)
Motifs rich in arginines and glycines were recognized several decades ago to play functional roles and were termed glycine-arginine-rich (GAR) domains and/or RGG boxes. We review here the evolving functions of the RGG box along with several sequence variations that
Ahmed Moursy et al.
Nucleic acids research, 42(10), 6659-6672 (2014-04-03)
Regulation of SMN2 exon 7 splicing is crucial for the production of active SMN protein and the survival of Spinal Muscular Atrophy (SMA) patients. One of the most efficient activators of exon 7 inclusion is hnRNP G, which is recruited
Diane N Singhroy et al.
Retrovirology, 10, 73-73 (2013-07-23)
Protein arginine methyltransferase 6 (PRMT6) is a nuclear enzyme that methylates arginine residues on histones and transcription factors. In addition, PRMT6 inhibits HIV-1 replication in cell culture by directly methylating and interfering with the functions of several HIV-1 proteins, i.e.
Jia-Yi Dong et al.
American heart journal, 162(6), 959-965 (2011-12-06)
Previous studies suggest that L-arginine, an amino acid and a substrate of nitric oxide synthase, may have blood pressure (BP)-lowering effect. Because some studies were performed with limited number of patients with hypertension and therefore limited statistical power with sometimes
Kaoru Kumazaki et al.
Nature, 509(7501), 516-520 (2014-04-18)
Newly synthesized membrane proteins must be accurately inserted into the membrane, folded and assembled for proper functioning. The protein YidC inserts its substrates into the membrane, thereby facilitating membrane protein assembly in bacteria; the homologous proteins Oxa1 and Alb3 have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service