Skip to Content
Merck
All Photos(2)

Key Documents

SFABIKB

Roche

KAPA SYBR® FAST

suitable for qPCR, 2 ×, ABI PRISM®

Synonym(s):

qPCR

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€104.00
5 ML
€447.00
10 ML
€816.00
50 ML
€2,540.00

€104.00


Please contact Customer Service for Availability


Select a Size

Change View
1 ML
€104.00
5 ML
€447.00
10 ML
€816.00
50 ML
€2,540.00

About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

€104.00


Please contact Customer Service for Availability

Quality Level

usage

sufficient for 100 reactions
sufficient for 1000 reactions
sufficient for 500 reactions
sufficient for 5000 reactions

shelf life

≤12 mo.

feature

dNTPs included
hotstart

packaging

kit of 1 mL (100 x 20 μL rxn; KK4603)
kit of 10 mL (1000 x 20 μL rxn; KK4605)
kit of 5 mL (500 x 20 μL rxn; KK4604)
kit of 50 mL (5000 x 20 μL rxn; KK4617)

manufacturer/tradename

Roche

concentration

2 ×

technique(s)

qPCR: suitable

input

purified DNA

compatibility

for use with Applied Biosystems® 5700
for use with Applied Biosystems® 7000
for use with Applied Biosystems® 7300
for use with Applied Biosystems® 7700
for use with Applied Biosystems® 7900HT
for use with StepOne
for use with StepOnePlus

detection method

SYBR® Green

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

KAPA SYBR® FAST qPCR Kits contain the first DNA polymerase engineered via directed evolution to be more tolerant of SYBR Green I dye inhibition.

The improved robustness, processivity, and speed of KAPA SYBR FAST qPCR Kits result in consistently high amplification efficiencies enabling more accurate relative quantification for gene expression analysis. KAPA SYBR FAST qPCR Kits, developed to perform optimally in stringent real-time PCR reaction conditions, exhibit dramatic improvements to signal-to-noise ratio (fluorescence), quantification cycle (Cq), linearity, and sensitivity.

Application

KAPA SYBR® FAST qPCR kit can be used for:
  • Gene expression[1]
  • Gene knockdown validation
  • Microarray validation
  • ChIP analysis
  • Low copy detection
  • Absolute quantification of NGS libraries
  • Real-time PCR[2][3][4]

Features and Benefits

Quantitate changes in gene expression more accurately
  • High reaction efficiency between 95 – 105% improves accuracy and reproducibility.
  • Unbiased efficiency across a wide range of GC contents and amplicon lengths.

Detect low copy and difficult targets consistently
  • Improved processivity results in earlier Cq scores.
  • Higher fluorescence detection across varying AT- and GC-rich targets.
  • Novel enzyme is resistant to SYBR® inhibition.

Complete real-time PCR runs in just 40 minutes
  • 55% shorter run times with fast cycling protocol.
  • Maintain high performance when switching from standard to fast protocols.

Quick Notes:
  • This kit contains an engineered enzyme optimized for qPCR using SYBR Green I dye chemistry.
  • The 2X master mix contains a proprietary buffer. Together with the novel enzyme, this improves amplification efficiency of both GC- and AT-rich targets.
  • 20 sec initial denaturation at 95°C is sufficient for enzyme activation. When working with complex templates, an initial denaturation of 3 min is recommended.
  • For 3-step cycling, use 20 sec for primer annealing and 1 sec for extension/data acquisition at 72°C.
  • Do not exceed 25 μL reaction volumes.

Quality

All kit components are subjected to stringent functional quality control, are free of detectable contaminating exo and endonuclease activity, and meet strict requirements with respect to DNA contamination.

Preparation Note

Always ensure that components have been fully thawed and thoroughly mixed before use. KAPA SYBR® FAST qPCR Master Mix (2X) may not freeze solidly, even when stored at -20°C.

The SYBR Green I dye contained in KAPA SYBR FAST qPCR Master Mix (2X) ABI PRISM® and ROX/fluorescein dyes (depending on kit configuration) are light sensitive. Exposure to direct light for an extended period of time will result in loss of fluorescent signal intensity.

KAPA SYBR FAST qPCR Master Mix (2X) is stable through 30 freeze-thaw cycles. Ensure that all reagents are stored protected from light at -20°C when not in use. When protected from light, reagents are stable in the dark at 4°C for at least one week and may be stored at this temperature for short-term use, provided that they do not become contaminated with microbes and/or nucleases.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

ABI PRISM is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Applied Biosystems is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
SYBR is a registered trademark of Life Technologies
StepOne is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
StepOnePlus is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries

Kit Components Only

Product No.
Description

  • KAPA SYBR® FAST DNA Polymerase

  • reaction buffer

  • dNTPs

  • SYBR® Green I dye

  • MgCl2 2.5 mM

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MicroRNA-138 suppresses ovarian cancer cell invasion and metastasis by targeting SOX4 and HIF-1a
Yu-Ming Yeh
International Journal of Cancer. Journal International Du Cancer, 133(4), 867-878 (2013)
CD1d induction in solid tumor cells by histone deacetylase inhibitors through inhibition of HDAC1/2 and activation of Sp1.
Yang Pei-Ming, et al.
Epigenetics, 7(4), 390-399 (2012)
Establishment of alternative culture method for spermatogonial stem cells using knockout serum replacement.
Aoshima K, et al.
PLoS ONE, 8(10), e77715-e77715 (2013)
SMN is required for the maintenance of embryonic stem cells and neuronal differentiation in mice.
Chang W F, et al.
Brain Structure &Amp; Function, 220(3), 1539-1553 (2015)
Pei-Ming Yang et al.
Epigenetics, 7(4), 390-399 (2012-03-16)
CD1d is a MHC class-like molecule that presents glycolipids to natural killer T (NKT) cells, then regulates innate and adaptive immunity. The regulation of CD1d gene expression in solid tumors is still largely unknown. Gene expression can be epigenetically regulated

Articles

An overview of directed evolution and the methods for generating proteins with optimized or entirely new functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service