Skip to Content
Merck
All Photos(1)

Key Documents

11498045910

Roche

Actin RNA Probe, DIG-labeled

greener alternative

solution, pkg of 2 μg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324

form

solution

Quality Level

packaging

pkg of 2 μg

manufacturer/tradename

Roche

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

General description

This antisense RNA probe (human β-actin) is in vitro transcribed in the presence of digoxigenin (DIG)-UTP. The transcript has a length of 588 bases. 550 bases are complementary to the 5′ region of human β-actin mRNA (nucleotides 69 to 6l8, EMBL: HSAC07). The additional 25 bases at the 5′ end, and the 13 additional bases at the 3′ end of the transcript are specific for the promoter/polylinker region of the transcription vector.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical.  The DIG System was established as a sensitive and cost-effective alternative to radioactivity for the labeling and detection of nucleic acids. There are many available publications that prove the versatility of the DIG System, so use of radio-labeling is no longer the only option for labeling of DNA for hybridization.

Application

Actin RNA Probe, DIG-labeled, is specifically useful for evaluating the quality and quantity of various RNA species and can be used:
  • in In situ hybridization (for example, as a control in mRNA detection)
  • for quality control in the construction of cDNA libraries
  • in northern blot analysis to evaluate RNA from various human cell lines and tissue samples

Components

Actin RNA Probe, DIG-labeled, is supplied in autoclaved, DEPC-treated water.

Sequence

Agarose gel electrophoresis under denaturing conditions and subsequent northern blot analysis reveal a defined band of 588 bases.

Preparation Note

Working solution: Dilute Sample
  • Remove the desired number of isotyping strips from the canister. Remove the caps from an equal number of development tubes.
    The tubes may be labeled with a pencil or felt-tipped lab marker for easy identification.
  • Dilute a sample containing the mouse monoclonal antibody in 1% BSA/ phosphate-buffered saline (PBS), pH 7.2 to 7.6.
    Culture supernatant samples should be diluted 1:10 to 1:100. Ascites samples should be diluted 1:20,000. These are recommended dilutions and may vary depending on the concentration of antibody in your sample. In our experience, a monoclonal antibody
    concentration of 0.1 to 1 g/ml of diluted sample gives the best results. 150 ml of this diluted sample will be added to the development tube.

Storage conditions (working solution): During assay, the preparation should be maintained at 0 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Tsuruga et al.
Journal of dental research, 81(11), 771-775 (2002-10-31)
The elastic system fibers consist of three types--oxytalan, elaunin, and elastic fibers--differing in their relative microfibril and elastin contents. All three types are found in human gingiva, but human periodontal ligaments contain only elastin-free fibers. We examined cultured human gingival
Saligrama R Kalpana et al.
Journal of infection in developing countries, 15(4), 566-572 (2021-05-07)
Acute Rheumatic Fever/ Rheumatic Heart Disease (ARF/RHD), a sequel of group A streptococcal (GAS) infection, even today constitutes a public health issue in developing countries including India. Differences in the prevalence of ARF/RHD in countries with a similar prevalence of

Articles

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service