Skip to Content
Merck
All Photos(1)

Documents

11371517001

Roche

Swa I

from Staphylococcus warneri

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

solution

packaging

pkg of 200 U (10 U/μl)

manufacturer/tradename

Roche

parameter

25 °C optimum reaction temp.

shipped in

dry ice

storage temp.

−20°C

Specificity

Recognition sites: ATTTAAAT
ATTTAAAT
Restriction site: ATTT↓AAAT
ATTT↓AAAT
Heat inactivation: Swa I can be heat inactivated by incubation at 65 °C for 20 minutes.

Application

Swa I is very useful for digesting GC-rich bacterial genomes.

DNA Profile

Number of cleavage sites on different DNAs
  • λ: 0
  • φX174: 0
  • Ad2: 1
  • M13mp7: 1
  • pBR322: 0
  • pBR328: 0
  • SV40: 1

Unit Definition

One unit is the enzyme activity that completely cleaves 1 μg Ad2 × Ssp I DNA in one hour at +25 °C in a total volume of 25 μl (1x) SuRE/Cut buffer H.

Analysis Note

SuRE/Cut Buffer System
The buffer in bold is recommended for optimal activity
  • A: 0-10%
  • B: 10-25%
  • H: 100%
  • L: 0-10%
  • M: 0-10%

Activity in PCR buffer: Not tested

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Enzyme Solution

  • SuRE/Cut Buffer H 10x concentrated

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service