Skip to Content
Merck
All Photos(1)

Key Documents

11093037001

Roche

Chromozym t-PA

powder, 90% (enzymatic), 0.25 mM

Synonym(s):

N-Methylsulfonyl-D-Phe-Gly-Arg-4-nitranilide acetate

Sign Into View Organizational & Contract Pricing

Select a Size

20 MG
€427.00

€427.00


Please contact Customer Service for Availability


Select a Size

Change View
20 MG
€427.00

About This Item

UNSPSC Code:
12352204

€427.00


Please contact Customer Service for Availability

Quality Level

Assay

90% (enzymatic)

form

powder

mol wt

636.7

packaging

pkg of 20 mg

manufacturer/tradename

Roche

concentration

0.25 mM

storage temp.

20-25°C

Related Categories

General description

N-Methylsulfonyl-D-Phe-Gly-Arg-4-nitranilide acetate

Application

Use Chromozym® t-PA (tissue plasminogen activator) (C24H32N8O7S x CH3COOH) as a substrate for the determination of t-PA, both in purified preparations and in cell culture supernatants. Moreover, it can be applied to evaluate the content of one-chain and two-chain t-PA.[1]

Quality

Purity: >90% (enzymatic)
Contaminant: <0.5% free 4-nitraniline

Sequence

N-Methylsulfonyl-D-Phe-Gly-Arg-4-nitranilide-acetate

Preparation Note

Working concentration: Approximately 0.25 mM
Storage conditions (working solution): 2 to 8 °C
An aqueous solution of Chromozym t-PA (4mM) is stable for at least two weeks if stored at 2 to 8 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

Chromozym is a registered trademark of Pentapharm AG, Basel

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucia Cilenti et al.
The Journal of biological chemistry, 278(13), 11489-11494 (2003-01-17)
Omi/HtrA2 is a mammalian serine protease with high homology to bacterial HtrA chaperones. Omi/HtrA2 is localized in mitochondria and is released to the cytoplasm in response to apoptotic stimuli. Omi/HtrA2 induces cell death in a caspase-dependent manner by interacting with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service