Skip to Content
Merck
All Photos(1)

Key Documents

10663573001

Roche

BM-Condimed

suitable for cell culture, solution, pkg of 100 mL

Synonym(s):

growth factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

sterility

0.2 μm filtered

form

solution

packaging

pkg of 100 mL

manufacturer/tradename

Roche

technique(s)

cell culture | mammalian: suitable

solubility

water: miscible

General description

BM-Condimed is prepared from the supernatant of human peripheral blood mononuclear cells stimulated with Con A. It contains a complex mixture of growth factors and lymphokines.

Application

BM-Condimed is used as a supplement (10%, v/v) to the normal culture basal medium (e.g., F 10, M 199, RPMI 1640, DMEM), containing 10 to 20% FCS for the cultivation of human amniotic and chorionic villi cells. BM-Condimed was also shown to improve culture conditions for bone marrow cells and peripheral blood leukocytes.

Physical form

Physiological solution, filtered through a 0.2 μm pore size membrane, buffered with sodium bicarbonate.

Preparation Note

Working concentration: 1 to 100μg/ml
1 to 100μg/ml
Working solution: Note: To 90ml of the normal basal medium F 10, M 199 Hanks, RPMI 1640, MEM Dulbecco, MEM Earle, Alpha MEM (containing 10 to 20% fetal bovine serum), add 10ml BM-Condimed. Do not use as a basal medium or as a replacement for serum. One percent L-glutamine may be added to the mixture.
Biological activity: Each lot is assayed for its ability to stimulate the colony growth of primary amniotic cells.
Source: BM-Condimed is prepared from the supernatant of human peripheral blood mononuclear cells stimulated with Con A. It contains a complex mixture of growth factors and lymphokines.

Note: Do not add antibiotics, as these are already present in BM-Condimed, and additional antibiotics will affect the growth of the cells.

Storage and Stability

Store at 2 to 8 °C. (Store in dark!)

Other Notes

For life science research only. Not for use in diagnostic procedures.

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vascular E-selectin Expression Detected in Formalin-fixed, Paraffin-embedded Sections With an E-selectin Monoclonal Antibody Correlates With Ulcerative Colitis Activity
Murahashi, et al.
The Journal of Histochemistry and Cytochemistry, 70 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service