Skip to Content
Merck
All Photos(1)

Key Documents

PRIH95E94E13

Princeton BioMolecular Research, Inc.

2-[2-(4-Amino-phenoxy)-ethoxy]-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
197.23
MDL number:

Iupac Name

2-[2-(4-aminophenoxy)ethoxy]ethanol

Inchi Code

1S/C10H15NO3/c11-9-1-3-10(4-2-9)14-8-7-13-6-5-12/h1-4,12H,5-8,11H2

InChI key

CJGYVGZZQMVWBL-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

Oil

Product Link

http://store.princetonbio.com/?product_id=PBMR051843

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library