PRIH93ED81FD
Princeton BioMolecular Research, Inc.
2'-(2-chlorophenyl)-1',2'-dihydro-3'H-spiro[cyclopentane-1,4'-isoquinolin]-3'-iminium bromide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
391.74
Iupac Name
2'-(2-chlorophenyl)-1'H-spiro[cyclopentane-1,4'-isoquinolin]-3'(2'H)-iminium bromide
Inchi Code
1S/C19H19ClN2.BrH/c20-16-9-3-4-10-17(16)22-13-14-7-1-2-8-15(14)19(18(22)21)11-5-6-12-19;/h1-4,7-10,21H,5-6,11-13H2;1H
InChI key
OXFSTVMYTQRRHB-UHFFFAOYSA-N
Purity
95%
Country of Origin
UA
Physical Form
Solid
Product Link
http://store.princetonbio.com/?product_id=PBMR362834
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.