PRIH11AAF2CA
Princeton BioMolecular Research, Inc.
4,7,7-trimethyl-2,3-dioxo-N-[4-(trifluoromethoxy)phenyl]bicyclo[2.2.1]heptane-1-carboxamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
369.34
MDL number:
Iupac Name
4,7,7-trimethyl-2,3-dioxo-N-[4-(trifluoromethoxy)phenyl]bicyclo[2.2.1]heptane-1-carboxamide
Inchi Code
1S/C18H18F3NO4/c1-15(2)16(3)8-9-17(15,13(24)12(16)23)14(25)22-10-4-6-11(7-5-10)26-18(19,20)21/h4-7H,8-9H2,1-3H3,(H,22,25)
InChI key
UXMAFAYQPNMMER-UHFFFAOYSA-N
Purity
95%
Country of Origin
UA
Physical Form
Solid
Product Link
http://store.princetonbio.com/?product_id=PBMR278545
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.