Key Documents
ZLXL00ESV
Milli-Q
External Solenoid valve
For use with Milli-Q® CLX / HR / HX systems
Select a Size
1 EA
€1,030.00
€1,030.00
Available to ship on11 April 2025Details
Request more information
Select a Size
Change View
1 EA
€1,030.00
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
€1,030.00
Available to ship on11 April 2025Details
Request more information
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
General description
The External Solenoid Valve is used to stop water flow downstream of its location if a leak is detected by the system.
Application
Used to protect the lab surroundings or other instruments from water damage, when used in conjunction with a water sensor.
Features and Benefits
Genuine replacement part for External Solenoid Valve for high-flow systems | ZLXL00ESV
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany