Skip to Content
Merck
All Photos(1)

Key Documents

ZFCBEQ0EU

Milli-Q

Country Box Milli-Q® EQ Systems

Available only in European Union

Synonym(s):

Country box Milli-Q® EQ 7000 EU

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€134.00

€134.00


Estimated to ship on14 April 2025Details



Select a Size

Change View
1 KIT
€134.00

About This Item

UNSPSC Code:
41104214
NACRES:
SB.51

€134.00


Estimated to ship on14 April 2025Details


packaging

pkg of 1 kit

compatibility

for use with Milli-Q® EQ 7000

Looking for similar products? Visit Product Comparison Guide

General description

The Country box for the Milli-Q® EQ 7000 system is an installation accessory that contains a printed quick reference guide, a user manual in English on a USB key, and a power cord adapted to the local electrical network.

Application

The components of the Country box assist users to quickly access the information they need to correctly use their system and connect to the local power network.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Available only in the Netherlands, Sweden, Poland, Finland, Norway, South Korea, United Arab Emirates, Israel, Czech Republic, Slovenia, Hungary, Vietnam, Saudi Arabia, Philippines, Iceland, Greece, Indonesia, Morocco, Turkey, Bangladesh, Lithuania, Slovakia, Ukraine, Egypt, Croatia

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service