Skip to Content
Merck
All Photos(3)

Key Documents

ZEQ7FTPDL

Milli-Q

Foot Pedal

Hands-free dispensing accessory for Milli-Q® EQ 7000 water purification systems

Synonym(s):

Milli-Q® EQ 7000 Foot Pedal

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€305.00

€305.00


Available to ship on15 April 2025Details


Request more information

Select a Size

Change View
1 EACH
€305.00

About This Item

UNSPSC Code:
41104200
NACRES:
JA.13

€305.00


Available to ship on15 April 2025Details


Request more information

feature

Remotely controls water delivery by a simple push of the foot.

packaging

pkg of 1 unit

manufacturer/tradename

Q-POD®

compatibility

for use with Milli-Q® EQ 7000

Looking for similar products? Visit Product Comparison Guide

General description

The foot pedal accessory connected to the Q-POD® dispenser allows to start and stop dispensing ultrapure (Type 1) water, by a simple push of the foot. When combined with Volumetric Dispense mode, one press on the pedal starts the production of ultrapure water at the point of dispense. Dispensing automatically stops at the set volume.

Application

This foot pedal is used for hands-free dispensing of ultrapure water at POD (point of dispense) of Milli-Q® EQ 7000 water systems.

Features and Benefits

  • Ideal for scientists who need their free hands when accessing purified water.
  • Volumetric dispense mode saves time when repeat dispensing the same volume.
  • Supports lab sanitary protocols as users do not need to touch the system to access water.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service