Key Documents
YY2229353
Millipore
PTFE O-ring 293-Disc Filter Holder
Select a Size
1 EA
€134.00
€134.00
Estimated to ship on31 July 2025Details
Select a Size
Change View
1 EA
€134.00
About This Item
UNSPSC Code:
41104909
eCl@ss:
32031613
€134.00
Estimated to ship on31 July 2025Details
material
PTFE
Quality Level
compatibility
for use with 293-mm Stainless Steel Sanitary Filter Holder
General description
The PTFE O-ring 293-Disc Filter Holder is intended for use with the 293 mm stainless steel sanitary filter holder - product number YY3029316 - an ideal solutions for sterilizing liquids or gases by pressure filtration.