Key Documents
XX6320022
Millipore
Battery Charger
for use with Millipore Single/Dual Chamber Incubator, AC/DC input 230 V, 50 Hz
Select a Size
1 EA
€296.00
€296.00
Estimated to ship on22 April 2025Details
Request more information
Select a Size
Change View
1 EA
€296.00
About This Item
UNSPSC Code:
41104424
€296.00
Estimated to ship on22 April 2025Details
Request more information
Quality Level
application(s)
bioburden testing
cosmetics
environmental
food and beverages
compatibility
for use with Millipore Single/Dual Chamber Incubator
General description
The Battery Charger for Single/Dual chamber incubator is designed to charge the rechargeable nickel- cadmium battery installed in the incubator.