Skip to Content
Merck
All Photos(1)

Key Documents

XX6320001

Millipore

Nickel-cadmium Battery

Beer, Bottled Water, Cosmetics, Pharmaceutical Products Analysis, Water Monitoring

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€595.00

€595.00


Estimated to ship on03 April 2026Details


Request more information

Select a Size

Change View
1 EA
€595.00

About This Item

UNSPSC Code:
41104424
NACRES:
NB.11

€595.00


Estimated to ship on03 April 2026Details


Request more information

input

sample type cosmetics
sample type bottled water
sample type beer
sample type pharmaceutical(s)

application(s)

cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

Incubate total and fecal coliforms, fecal streptococci and other microorganisms
- Complies with Standard Methods requirements
- View samples through clear plastic inner door without opening, preventing temperature fluctuations
- Compact lightweight portable unit for field use

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service