Skip to Content
Merck
All Photos(1)

Documents

XX20047RK

Millipore

Replacement parts kit, Hydrosol holder

Synonym(s):

Replacement Parts Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24

material

Nylon wheel (locking)
PTFE gasket
anodized aluminum lock ring

Quality Level

shipped in

ambient

General description

The Hydrosol filter holder is designed for the filtration of flammable liquids. Unlike other filter holders, the Hydrosol filter holder includes a grounding unit to prevent sparking from static discharge, which can be a safety hazard.

Applications:
Vacuum Filtration of Flammable Liquids

XX2004720 Hydrosol Filter Holder, 47 mm, stainless steel

Replacement Parts
XX2004704 Funnel and Locking Ring Assembly, stainless steel
XX2004708 Support Screen, 47 mm, stainless steel
XX2004702 Base with Screen, gasket and support
XX2004703 Gasket, PTFE
XX2004718 No. 8 Stopper, 9.5 mm (3/8 in.) hole, silicone
XX20047RK Replacement Parts Kit for Hydrosol Filter Holder

Accessories
XX1004705 Vacuum Filtering Flask, 1 L
XX1004744 Vacuum Filtering Flask, 4L
XX7100004 Tubing 3/16 in. I.D. x 140 cm, silicone
XX6200006P Filter Forceps, blunt end, stainless steel
XX2604735 3-place Manifold, 47 mm, PVC
XX2504735 3-place Manifold, 47 mm, stainless steel
XX2504700 6-place Manifold, 47 mm, stainless steel
WP6211560 High Output Pump, 115 V/60 Hz
WP6222050 High Output Pump, 220 V/50 Hz
WP6210060 High Output Pump, 100 V/50-60 Hz

Application

Accessories for filter holders for sample preparation.

Components

(1) 47mm locking ring, (1) hex key (1) set of nylon locking wheels and wrench (5) PTFE screen gaskets (5) PTFE lock ring gaskets

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service