Skip to Content
Merck
All Photos(1)

Key Documents

XT1200000

Millipore

Tubing for solvent/buffer, PTFE, 70 cm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€164.00

€164.00


Estimated to ship on28 May 2025Details



Select a Size

Change View
1 EA
€164.00

About This Item

UNSPSC Code:
41105101
eCl@ss:
32012303
NACRES:
NB.24

€164.00


Estimated to ship on28 May 2025Details


Agency

suitable for EPA OTM-45

Quality Level

manufacturer/tradename

MilliSolve

application(s)

PFAS testing

shipped in

ambient

General description

XT1200000 is a replacement part for the MilliSolve Filtration System.

The MilliSolve Filtration System has been designed for the filtration of liquid chromatography buffers and solvents under vacuum. The system uses a 0.45 µm membrane filter to eliminate particles which can shorten column life. Vacuum filtration with the MilliSolve system also removes a large portion of dissolved gases from buffers, reducing the risk of air bubbles interfering with LC instruments. With automatic and continuous filtration, there is no need to add liquid into a funnel as filtration progresses. Filtration occurs in a closed system, which is important when filtering hazardous fluids. If open funnel filtration is required, the 300 mL funnel replaces the vacuum cover.

Features & Benefits:
•Automatic, continuous solvent and buffer filtration eliminates the need to pour during filtration
•2 L receiving flask includes plug and tubing for storage, eliminating filtered buffer transfer, reducing labor and lessening contamination risk
•Conical 2 L flask bottom allows access to all filtered solvent or buffer

Applications:
Mobile Phase Preparation, Buffer Filtration

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service