WBLUR0020
Immobilon® Crescendo Western HRP substrate
Western blotting, dot/slot blotting
About This Item
Recommended Products
product name
Immobilon Crescendo Western HRP substrate, 20 mL, Western blotting, dot/slot blotting
conjugate
peroxidase conjugate
Quality Level
packaging
pkg of 100 mL
manufacturer/tradename
Immobilon®
technique(s)
dot blot: suitable
western blot: suitable
membrane area
200 cm2 , membrane coverage
detection method
chemiluminescent
shipped in
ambient
storage temp.
2-8°C
General description
Application
Packaging
Quality
Storage and Stability
Legal Information
Disclaimer
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A discussion of factors affecting observed vs. calculated molecular weight in gel electrophoresis and western blotting.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service