Skip to Content
Merck
All Photos(1)

Key Documents

VSWP29325

Millipore

MF-Millipore Membrane

pore size 0.025 μm, filter diam. 293 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane, mixed cellulose esters, Hydrophilic, 0.025 μm, 293 mm, white, plain

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€1,535.00

€1,535.00


Estimated to ship on14 April 2025Details



Select a Size

Change View
25 EA
€1,535.00

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602

€1,535.00


Estimated to ship on14 April 2025Details


material

mixed cellulose esters (MCE)
plain filter
white filter

Quality Level

description

Microdialysis of DNA and proteins

feature

hydrophilic

manufacturer/tradename

MF-Millipore

parameter

0.15 L/min-cm2 air flow rate
0.15 mL/min-cm2 water flow rate
75 °C max. temp.

filter diam.

293 mm

thickness

105 μm

gravimetric extractables

1.5%

refractive index

n/D 1.5

matrix

MF-Millipore

pore size

0.025 μm pore size
72 % porosity

bubble point

≥21.1 bar, air with water at 23 °C

Looking for similar products? Visit Product Comparison Guide

General description

Filter Code: VSWP

Application

Microdialysis of DNA and proteins

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: BioProcessing
  • Intended Use: Reduction or removal of microorganism/bioburden
  • Instructions for Use: Please refer first page of Visual Inspection Guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

EU REACH Annex XIV (Authorisation List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service