Skip to Content
Merck

VPPH103NB1

Millipore

Viresolve® Pro Shield H

Virus prefilter, Viresolve® Pro Shield H, Modus 1.3 format, filtration area 0.22 m²

Synonym(s):

Viresolve® Pro Shield H Modus 1.3

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€2,558.00

€2,558.00


Estimated to ship on02 June 2025Details



Select a Size

Change View
1 EA
€2,558.00

About This Item

UNSPSC Code:
23151806
eCl@ss:
32031690

€2,558.00


Estimated to ship on02 June 2025Details


material

PVDF housing
polyethersulfone membrane
silicone O-ring
white device

Quality Level

product line

EMPROVE® Filter

manufacturer/tradename

Viresolve®

parameter

4.1 bar max. inlet pressure (60 psig)

H

13.56 cm (5.34 in.)

L × W × H

13.6 cm (5.3 in.) × 18.2 cm (7.3 in.) × 9.2 cm (3.6 in.)

L

18.62 cm (7.33 in.)

W

9.22 cm (3.63 in.)

filtration area

0.22 m2

membrane nominal area

0.22 m2

weight

1.39 kg (3.06 lb)

impurities

<0.25 EU/mL USP bacterial endotoxins (LAL test, sample aqueous extraction)

fitting

3/4 in. inlet/outlet sanitary fitting(s)
1/8 in. integrated vent hose barb (with double O-ring seal)

Looking for similar products? Visit Product Comparison Guide

General description

The Viresolve® Pro Solution provides a comprehensive, flexible template solution for viral clearance within mAb and recombinant protein bioproduction. This proven viral clearance solution delivers the highest levels of retention assurance and productivity across a broad range of feed stream characteristics.

Viresolve Pro Modus Devices are typically used for filtration of pilot to mid-scale batch volumes of proteins. The Viresolve® Pro Shield H Modus 1.3 offers a filtration area of 0.22 m2 and has a primary use in pilot/small-volume processing.

Features and Benefits

  • Pilot and mid scale manufacturing
  • Three different size formats
  • Fully self contained (hardware holder not required)
  • Fully disposable single-use fluid path
  • Devices 100% tested with air/water diffusion and Binary Gas Test
  • Shield / Shield H 100% aerosol tested

Other Notes

These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices.

Preparation Note

This product was manufactured with materials that meet the criteria for a "non-fiber releasing" filter as defined in 21 CFR 210.3 (b) (6).

Storage and Stability

4°C - 30°C

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
VIRESOLVE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

EMD Millipore Corporation certifies that this product complies with the European Pressure Equipment Directive, 97/23/EC of 29 May 1997. This product has been classified under Article 3 § 3 of the Pressure Vessel Directive. It has been designed and manufactured in accordance with sound engineering practice to ensure safe use. In compliance with Article 3 § 3 of this Pressure Vessel Directive, this product does not bear the CE mark.

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service