Skip to Content
Merck
All Photos(2)

Key Documents

UFSC20001

Amicon® Stirred Cell 200mL

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Synonym(s):

Amicon Cell, Stirred Cell, Stirred Cell 200mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€1,910.00

€1,910.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
1 EA
€1,910.00

About This Item

UNSPSC Code:
41104903
eCl@ss:
32031690

€1,910.00


Estimated to ship on22 April 2025Details


material

316 stainless steel ring(s) (stir bar retaining)
316 stainless steel washer
Nylon connections (Luer on filtrate tubing)
Tygon® tubing (filtrate; E-3603)
polyacetal base
polyacetal support (for stir bar)
polyethylene tubing (pressure)
polysulfone body
polysulfone cap
polysulfone holder (for membrane)
polysulfone stir bar(s)
silicone O-ring
silicone gasket

Quality Level

product line

Amicon®

manufacturer/tradename

Amicon®

parameter

200 mL process volume

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

H

15.5 cm (6.1 in.)

filter diam.

63.5 mm

filtration area

28.7 cm2

shipped in

ambient

Application

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Principle

Pressure

Linkage

Replaces: 5123

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Questions

  1. Can the Amicon Stirred Cell Units be autoclaved?

    1 answer
    1. The general user instructions for the Stirred Cell units (UFSC05001 - 50 ml, UFSC20001 - 200 ml, UFSC40001 - 400 ml) can be found online. The Stirred Cell units can be autoclaved for at least 10 cycles at 121 °C, 1 bar (250 °F, 15 psi) for 30 minutes, with a slow exhaust cycle. However, it's important to note that due to variables beyond our control, no warranty is provided or implied for more than 10 autoclave cycles. Additionally, the pressure inlet tubing assembly is NOT autoclavable. Other methods of sterilization/sanitization include sterilizing gases, 70% ethanol or isopropanol for sanitization, and 5% formalin for disinfection.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service